Bromocresol green in ethanol (contains NaOH) stain, for TLC

Supplier: TCI America

Synonyms: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

B2401-500ML
TCB2401-500MLEA 144.49 USD
TCB2401-500ML
Bromocresol green in ethanol (contains NaOH) stain, for TLC
Bromocresol green

[for TLC Stain]
CAS Number: 76-60-8
MDL Number: MFCD00005874
Molecular Formula: C21H14Br4O5S
Molecular Weight: 698.01
Form: Liquid
Color: Deep Yellow Green
Flash Point (°C): 13

Formula: C₂₁H₁₄Br₄O₅S
MW: 698.02 g/mol
Flash Pt: 13 °C
MDL Number: MFCD00005874
CAS Number: 76-60-8
EINECS: 263-657-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=feea336c27abb6b584c4a46410bbebb0051f875a&vid=6bdf6fed94e2d3ef5c33d1afa7d034b379e7184b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR