Benzylchlorodimethylsilane ≥98.0% (by GC)

Supplier: TCI America

B2334-5G
TCB2334-5GEA 196.38 USD
TCB2334-5G
Benzylchlorodimethylsilane ≥98.0% (by GC)
Benzylchlorodimethylsilane

CAS Number: 1833-31-4 MDL Number: MFCD00018089 Molecular Formula: C9H13ClSi Molecular Weight: 184.74 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 211 Flash Point (°C): 73 Specific Gravity (20/20): 1.02

Formula: C₉H₁₃ClSi
MW: 184.74 g/mol
Boiling Pt: 211 °C
Density: 1.02 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00018089
CAS Number: 1833-31-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70897ac32445028e88abb5e5bb94d81b5d65a7e6&vid=70897ac32445028e88abb5e5bb94d81b5d65a7e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR