(+/-)-ɑ-Bisabolol ≥90.0%

Supplier: TCI America

Synonyms: rel-(R)-6-Methyl-2-((R)-4-methylcyclohex-3-en-1-yl)hept-5-en-2-ol, 6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol

B2119-25ML
TCB2119-025MLEA 86.8 USD
TCB2119-025ML
(+/-)-ɑ-Bisabolol ≥90.0%
(+/-)-alpha-Bisabolol

CAS Number: 515-69-5 MDL Number: MFCD00129082 Molecular Formula: C15H26O Molecular Weight: 222.37 Purity/Analysis Method: 90.0% (GC) Form: Clear Liquid Flash Point (°C): 135 Specific Gravity (20/20): 0.93


Product Categories: Bioscience, Sesquiterpenes Other Product Names / Synonyms: 6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol

Formula: C₁₅H₂₆O
MW: 222.37 g/mol
Density: 0.93 (20 °C)
Flash Pt: 135 °C
MDL Number: MFCD00129082
CAS Number: 515-69-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eaac698f97c29f3d4bea9e46fa0c9b4911db833b&vid=eaac698f97c29f3d4bea9e46fa0c9b4911db833b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR