2-Bromo-4-methylpyridine ≥98.0%

Supplier: TCI America

Synonyms: 2-Bromo-4-picoline

B2112-5G B2112-25G
TCB2112-005GEA 75.3 USD
TCB2112-005G TCB2112-25G
2-Bromo-4-methylpyridine ≥98.0%
2-Bromo-4-methylpyridine

CAS Number: 4926-28-7 MDL Number: MFCD00082590 Molecular Formula: C6H6BrN Molecular Weight: 172.03 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Flash Point (°C): 110 Specific Gravity (20/20): 1.53


Other Product Names / Synonyms: 2-Bromo-4-picoline

Formula: C₆H₆BrN
Boiling Pt: 42…44 °C (0.6 mmHg)
Melting Pt: 218…228 °C
Density: 1.53 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00082590
CAS Number: 4926-28-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d599944501be0af10873922aeceb9fcb06ab6a6&vid=3d599944501be0af10873922aeceb9fcb06ab6a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR