Trihexyl borate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 5337-36-0
MDL Number: MFCD00027295
Molecular Formula: C18H39BO3
Molecular Weight: 314.32
Purity/Analysis Method:
Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Boric Acid Trihexyl Ester
Formula:
C₁₈H₃₉BO₃ Boiling Pt: 311 °C Density: 0.85 (20 °C) Flash Pt: 149 °C |
MDL Number:
MFCD00027295 CAS Number: 5337-36-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...