3-(2-Benzothiazolyl)-7-(diethylamino)coumarin ≥98.0%
Supplier: TCI America
Synonyms:
3-(Benzo[d]thiazol-2-yl)-7-(diethylamino)-2H-chromen-2-one, Coumarin 6
B2088-1G
B2088-5G
TCB2088-1GEA
59.6
USD
TCB2088-1G
TCB2088-5G
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin ≥98.0%
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin
CAS Number: 38215-36-0
MDL Number: MFCD00041869
Molecular Formula: C20H18N2O2S
Molecular Weight: 350.44
Purity/Analysis Method:
Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Coumarin Dyes (Functional Organic Dyes)
Other Product Names / Synonyms: Coumarin 6
Formula:
C₂₀H₁₈N₂O₂S Melting Pt: 208 °C |
MDL Number:
MFCD00041869 CAS Number: 38215-36-0 EINECS: 253-830-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...