3-(2-Benzothiazolyl)-7-(diethylamino)coumarin ≥98.0%

Supplier: TCI America

Synonyms: 3-(Benzo[d]thiazol-2-yl)-7-(diethylamino)-2H-chromen-2-one, Coumarin 6

B2088-1G B2088-5G
TCB2088-1GEA 59.6 USD
TCB2088-1G TCB2088-5G
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin ≥98.0%
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin

CAS Number: 38215-36-0 MDL Number: MFCD00041869 Molecular Formula: C20H18N2O2S Molecular Weight: 350.44 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 208


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Coumarin Dyes (Functional Organic Dyes) Other Product Names / Synonyms: Coumarin 6

Formula: C₂₀H₁₈N₂O₂S
Melting Pt: 208 °C
MDL Number: MFCD00041869
CAS Number: 38215-36-0
EINECS: 253-830-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0b3221a5c293a551d900bf62c121ca0ec35ffe1&vid=b0b3221a5c293a551d900bf62c121ca0ec35ffe1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR