5-Bromo-2-iodotoluene ≥98.0%

Supplier: TCI America

B2069-5G B2069-25G
TCB2069-005GEA 29 USD
TCB2069-005G TCB2069-025G
5-Bromo-2-iodotoluene ≥98.0%
5-Bromo-2-iodotoluene

CAS Number: 116632-39-4 MDL Number: MFCD00060664 Molecular Formula: C7H6BrI Molecular Weight: 296.93 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 260 Flash Point (°C): 110 Specific Gravity (20/20): 2.10

Formula: C₇H₆BrI
Boiling Pt: 260 °C
Density: 2.10 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00060664
CAS Number: 116632-39-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6165b3c51ffa33eae8a77f7b5380dd54b89f492b&vid=6165b3c51ffa33eae8a77f7b5380dd54b89f492b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR