4-tert-Butylbenzenesulfonyl chloride ≥98.0%

Supplier: TCI America

Synonyms: 4-tert-Butylbenzene-1-sulfonyl chloride, 4-tert-Butylbenzene-1-sulphonyl chloride

B1983-25G B1983-5G
TCB1983-25GEA 94.2 USD
TCB1983-25G TCB1983-5G
4-tert-Butylbenzenesulfonyl chloride ≥98.0%
4-tert-Butylbenzenesulphonyl chloride

CAS Number: 15084-51-2 MDL Number: MFCD00007449 Molecular Formula: C10H13ClO2S Molecular Weight: 232.72 Purity/Analysis Method: 98.0% (T) Form: Crystal Boiling point (°C): 165 Melting point (°C): 81 Flash Point (°C): 110

Formula: C₁₀H₁₃ClO₂S
MW: 232.73
Boiling Pt: 165
Melting Pt: 83
Flash Pt: 110 °C
MDL Number: MFCD00007449
CAS Number: 15084-51-2
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR