Bis(pinacolato)diboron ≥99.0%

Supplier: TCI America

Synonyms: 4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane), 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane

B1964-5G B1964-1G B1964-25G B1964-100G
TCB1964-5GEA 97.91 USD
TCB1964-5G TCB1964-001G TCB1964-25G TCB1964-100G
Bis(pinacolato)diboron ≥99.0%
Bis(pinacolato)diboron

CAS Number: 73183-34-3 MDL Number: MFCD00799570 Molecular Formula: C12H24B2O4 Molecular Weight: 253.94 Purity/Analysis Method: 99.0% (GC) Form: Crystal Melting point (°C): 138


Product Categories: Synthetic Organic Chemistry, Suzuki-Miyaura Cross-coupling Reaction, Boration Other Product Names / Synonyms: 4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi-1,3,2-dioxaborolane

Formula: C₁₂H₂₄B₂O₄
MW: 253.94 g/mol
Melting Pt: 138 °C
MDL Number: MFCD00799570
CAS Number: 73183-34-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adea3dd4ce11ed26ddd715d93151dfc4ca405d9c&vid=adea3dd4ce11ed26ddd715d93151dfc4ca405d9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR