N-Boc-L-alaninol ≥98.0%
Supplier: TCI America
Synonyms:
Boc-L-Alaninol, Boc-Alaninol, (S)-2-(BOC-amino)-1-propanol
B1953-1G
B1953-5G
TCB1953-1GEA
27
USD
TCB1953-1G
TCB1953-005G
N-Boc-L-alaninol ≥98.0%
N-Boc-L-alaninol
CAS Number: 79069-13-9
MDL Number: MFCD00043121
Molecular Formula: C8H17NO3
Molecular Weight: 175.23
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: N-Boc-L-alaninol, (S)-2-(tert-Butoxycarbonylamino)-1-propanol
Formula:
C₈H₁₇NO₃ Melting Pt: 60 °C |
MDL Number:
MFCD00043121 CAS Number: 79069-13-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...