4-Bromobenzophenone ≥98.0%
Supplier: TCI America
B1917-25G
TCB1917-025GEA
70
USD
TCB1917-025G
4-Bromobenzophenone ≥98.0%
4-Bromobenzophenone
CAS Number: 90-90-4
MDL Number: MFCD00000103
Molecular Formula: C13H9BrO
Molecular Weight: 261.12
Purity/Analysis Method:
Formula:
C₁₃H₉BrO Boiling Pt: 176 °C Melting Pt: 82 °C |
MDL Number:
MFCD00000103 CAS Number: 90-90-4 EINECS: 202-024-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...