2,2'-Bipyridine-4,4'-dicarboxylic acid ≥96.0%
Supplier: TCI America
Synonyms:
4,4'-Dicarboxy-2,2'-bipyridine, [2,2'-Bipyridine]-4,4'-dicarboxylic acid
B1876-1G
B1876-100MG
TCB1876-1GEA
56
USD
TCB1876-1G
TCB1876-100MG
2,2'-Bipyridine-4,4'-dicarboxylic acid ≥96.0%
2,2'-Bipyridine-4,4'-dicarboxylic acid
CAS Number: 6813-38-3
MDL Number: MFCD00015430
Molecular Formula: C12H8N2O4
Molecular Weight: 244.21
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents, Solar Cell Research Reagents
Other Product Names / Synonyms: 2,2'-Biisonicotinic Acid
Formula:
C₁₂H₈N₂O₄ |
MDL Number:
MFCD00015430 CAS Number: 6813-38-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...