4-Bromomethyl-2-cyanobiphenyl ≥98.0%
Supplier: TCI America
Synonyms:
4'-(Bromomethyl)-[1,1'-biphenyl]-2-carbonitrile
B1806-25G
B1806-5G
TCB1806-025GEA
110
USD
TCB1806-025G
TCB1806-005G
4-Bromomethyl-2-cyanobiphenyl ≥98.0%
4-Bromomethyl-2-cyanobiphenyl
CAS Number: 114772-54-2
MDL Number: MFCD00671503
Molecular Formula: C14H10BrN
Molecular Weight: 272.15
Purity/Analysis Method:
Other Product Names / Synonyms: 4'-Bromomethylbiphenyl-2-carbonitrile, 2-Cyano-4'-bromomethylbiphenyl
Formula:
C₁₄H₁₀BrN MW: 272.14 g/mol Melting Pt: 127 °C |
MDL Number:
MFCD00671503 CAS Number: 114772-54-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...