Fullerene C70 ≥98.0% (by HPLC)

Supplier: TCI America

B1694-100MG
TCB1694-100MGEA 171 USD
TCB1694-100MG
Fullerene C70 ≥98.0% (by HPLC)
Fullerene C70

CAS Number: 115383-22-7
MDL Number: MFCD00146976
Molecular Formula: C70
Molecular Weight: 840.77
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Materials Chemistry, Organic Transistor Materials, Molecular Conductors, Solar Cell Research Reagents, Fullerenes (Nanocarbon Materials)
Other Product Names / Synonyms: Buckminsterfullerene C70, C70, Carbon C70

Formula: C₇₀
Melting Pt: > 280 °C
Density: 1.73 g/cm³ (20 °C)
MDL Number: MFCD00282904
CAS Number: 115383-22-7
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89d2c6a228d3a97aa6ff8f3c0b2bb5a1a40eb22e&vid=74702115f70659883415445e0bd7267e96d767b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR