Bis(triphenylphosphine)palladium(II) chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Bis(triphenylphosphine)palladium(II) dichloride
B1667-5G
B1667-25G
B1667-1G
TCB1667-005GEA
266.8
USD
TCB1667-005G
TCB1667-25G
TCB1667-001G
Bis(triphenylphosphine)palladium(II) chloride ≥98.0% (by titrimetric analysis)
Bis(triphenylphosphine)palladium (II) chloride
CAS Number: 13965-03-2
MDL Number: MFCD00009593
Molecular Formula: C36H30Cl2P2Pd
Molecular Weight: 701.90
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts, Metal Catalysts ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Dichlorobis(triphenylphosphine)palladium(II)
Formula:
C₃₆H₃₀Cl₂P₂Pd |
MDL Number:
MFCD00009593 CAS Number: 13965-03-2 EINECS: 237-744-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...