5-Bromo-2'-deoxyuridine (broxuridine) ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 59-14-3
MDL Number: MFCD00006529
Molecular Formula: C9H11BrN2O5
Molecular Weight: 307.10
Purity/Analysis Method:
Product Categories: Bioscience, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs
Other Product Names / Synonyms: 5-Bromo-1-(2-deoxy-beta-D-ribofuranosyl)uracil, 5-BrdU, BUdR
Formula:
C₉H₁₁BrN₂O₅ |
MDL Number:
MFCD00006529 CAS Number: 59-14-3 EINECS: 200-415-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...