N,N'-Bis(2-aminoethyl)-1,3-propanediamine ≥97.0%

Supplier: TCI America

Synonyms: 3,7-Diaza-1,9-nonanediamine, N1,N1'-(Propane-1,3-diyl)bis(ethane-1,2-diamine), 1,4,8,11-Tetraazaundecane

B1445-5G B1445-1G
TCB1445-005GEA 302 USD
TCB1445-005G TCB1445-1G
N,N'-Bis(2-aminoethyl)-1,3-propanediamine ≥97.0%
N,N'-Bis(2-aminoethyl)-1,3-propanediamine

CAS Number: 4741-99-5 MDL Number: MFCD00008174 Molecular Formula: C7H20N4 Molecular Weight: 160.27 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 287 Flash Point (°C): 110 Specific Gravity (20/20): 0.97


Product Categories: Materials Chemistry, Monomers, Bioscience, Polyamines Other Product Names / Synonyms: 3,7-Diaza-1,9-nonanediamine

Formula: C₇H₂₀N₄
Boiling Pt: 287 °C
Density: 0.97 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008174
CAS Number: 4741-99-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bce14eb64e0832fb6e65654c488b6ba8d9bcb2de&vid=bce14eb64e0832fb6e65654c488b6ba8d9bcb2de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR