Bisabolene (so called)
Supplier: TCI America
Synonyms:
4-(1,5-Dimethyl-4-hexenylidene)-1-methylcyclohexene
B1413-25ML
TCB1413-025MLEA
84
USD
TCB1413-025ML
Bisabolene (so called)
Bisabolene
[for Fragrance]
MDL Number: MFCD00129080
Molecular Formula: C15H24
Molecular Weight: 204.36
Form: Clear Liquid
Color: Slightly Pale Yellow
Flash Point (°C): 200
Formula:
C₁₅H₂₄ MW: 204.36 g/mol Flash Pt: 200 °C |
MDL Number:
MFCD00129080 CAS Number: 495-62-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...