Coomassie brilliant blue G-250

Supplier: TCI America

Synonyms: Brilliant blue G-250, Serva Blue G, Coomassie Blue G 250, Xylene Brilliant Cyanine G, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]-2'-methylbenzhydrylene]-3-methylcyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulfonatobenzyl)ammonium monosodium salt, Brillant blue G, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]-2'-methylbenzhydrylene]-3-methylcyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium salt, C.I. 42655, Acid Blue 90, Sodium 3-(((4-((4-((4-ethoxyphenyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)-2-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-methylphenyl)(ethyl)amino)methyl)benzenesulphonate, Brilliant indocyanine G, Brilliant Blue G, Sodium 3-(((4-((4-((4-ethoxyphenyl)amino)phenyl)(4-(ethyl(3-sulfonatobenzyl)iminio)-2-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-methylphenyl)(ethyl)amino)methyl)benzenesulfonate

B1146-25G
TCB1146-025GEA 51.11 USD
TCB1146-025G
Coomassie brilliant blue G-250
Coomassie brilliant blue G-250

CAS Number: 6104-58-1 MDL Number: MFCD00078482 Molecular Formula: C47H48N3NaO7S2 Molecular Weight: 854.02 Form: Crystal Color: Deep Red


Other Product Names / Synonyms: Acid Blue 90

Formula: C₄₇H₄₈N₃NaO₇S₂
MW: 854.04 g/mol
Density: >1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078482
CAS Number: 6104-58-1
EINECS: 228-058-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bc6c4eb62cd3d83c3603c41c8300585ed88e748&vid=0bc6c4eb62cd3d83c3603c41c8300585ed88e748&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR