Bromotrimethylsilane ≥95.0%

Supplier: TCI America

Synonyms: Trimethylbromosilane, Trimethylsilyl bromide

B1087-250ML B1087-25ML B1087-5ML
TCB1087-250MLEA 453 USD
TCB1087-250ML TCB1087-25ML TCB1087-5ML
Bromotrimethylsilane ≥95.0%
Bromotrimethylsilane

CAS Number: 2857-97-8 MDL Number: MFCD00000048 Molecular Formula: C3H9BrSi Molecular Weight: 153.09 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Boiling point (°C): 79 Melting point (°C): -43 Flash Point (°C): 32 Specific Gravity (20/20): 1.18


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Silicon Compounds (for Synthesis) Other Product Names / Synonyms: Trimethylbromosilane, Trimethylsilyl Bromide

Formula: C₃H₉BrSi
MW: 153.09 g/mol
Boiling Pt: 79
Melting Pt: -43
Density: 1.18 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00000048
CAS Number: 2857-97-8
UN: 2924
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6c574185fa8624d047a993a161724027e4da8c6&vid=c6c574185fa8624d047a993a161724027e4da8c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR