Chloro(1,5-cyclooctadiene)rhodium(I) dimer ≥98.0%
Supplier: TCI America
Synonyms:
(1,5-Cyclooctadiene)rhodium chloride dimer
CAS Number: 12092-47-6
MDL Number: MFCD00012415
Molecular Formula: C16H24Cl2Rh2
Molecular Weight: 493.08
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, C-H Bond Activation Reaction, Metal Catalysts ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Bis(1,5-cyclooctadiene)dirhodium(I) Dichloride, 1,5-Cyclooctadiene Rhodium(I) Chloride Dimer, [Rh(COD)Cl]2
Formula:
C₁₆H₂₄Cl₂Rh₂ MW: 493.08 g/mol |
MDL Number:
MFCD00012415 CAS Number: 12092-47-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...