2-Bromo-1-ethylpyridinium tetrafluoroborate ≥98.0%
Supplier: TCI America
Synonyms:
BEP, 2-Bromo-1-ethyl-pyridinium tetrafluoroborate, 2-Bromo-1-ethylpyridin-1-ium tetrafluoroborate
B1036-5G
B1036-25G
TCB1036-005GEA
94
USD
TCB1036-005G
TCB1036-025G
2-Bromo-1-ethylpyridinium tetrafluoroborate ≥98.0%
2-Bromo-1-ethylpyridinium tetrafluoroborate
CAS Number: 878-23-9
MDL Number: MFCD00060046
Molecular Formula: C7H9BBrF4N
Molecular Weight: 273.86
Purity/Analysis Method:
Formula:
C₇H₉BBrF₄N Melting Pt: 103 °C |
MDL Number:
MFCD00060046 CAS Number: 878-23-9 EINECS: 212-900-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...