5-Bromo-2-furoic acid ≥98.0%

Supplier: TCI America

Synonyms: 5-Bromofuran-2-carboxylic acid, 5-Bromofuroic acid

B1008-25G
TCB1008-025GEA 39.8 USD
TCB1008-025G
5-Bromo-2-furoic acid ≥98.0%
5-Bromo-2-furoic acid

CAS Number: 585-70-6 MDL Number: MFCD00003239 Molecular Formula: C5H3BrO3 Molecular Weight: 190.98 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 189


Other Product Names / Synonyms: 5-Bromo-2-furancarboxylic Acid, 5-Bromopyromucic Acid

Formula: C₅H₃BrO₃
MW: 190.98 g/mol
Melting Pt: 189 °C
MDL Number: MFCD00003239
CAS Number: 585-70-6
EINECS: 209-559-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13efc3d941d93ca30c086d37ab7c690825fef98b&vid=13efc3d941d93ca30c086d37ab7c690825fef98b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR