3,3',4,4'-Benzophenonetetracarboxylic dianhydride ≥96.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 2421-28-5
MDL Number: MFCD00005923
Molecular Formula: C17H6O7
Molecular Weight: 322.23
Purity/Analysis Method:
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Benzophenones (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Carbonyldiphthalic Anhydride
Formula:
C₁₇H₆O₇ Melting Pt: 226 °C Flash Pt: 324 °C |
MDL Number:
MFCD00005923 CAS Number: 2421-28-5 EINECS: 219-348-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...