Brucine, anhydrous ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2,3-Dimethoxystrychnidin-10-one, 10,11-Dimethoxystrychnine

B0946-25G
TCB0946-025GEA 83 USD
TCB0946-025G
Brucine, anhydrous ≥98.0% (by HPLC, titration analysis)
Brucine

CAS Number: 357-57-3
MDL Number: MFCD00005942
Molecular Formula: C23H26N2O4
Molecular Weight: 394.47
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 179


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Bioscience, Alkaloids (Phytochemicals), Indole Alkaloids
Other Product Names / Synonyms: 2,3-Dimethoxystrychnidin-10-one, 10,11-Dimethoxystrychnine

Formula: C₂₃H₂₆N₂O₄
MW: 394.47 g/mol
Melting Pt: 179 °C
MDL Number: MFCD00005942
CAS Number: 357-57-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4468ebc878ce0282d213bf13084a072d3e68061&vid=b0dd0ada8d80d00baa55bc5caaa30b2aa3d419ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR