1-Butanol ≥99.0%

Supplier: TCI America
Danger

Synonyms: Propylcarbinol, Methylolpropane, Butan-1-ol, Butyl alcohol, n-Butyl alcohol, Butyl hydroxide, 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxybutane, 1-Hydroxy butane, n-Butan-1-ol, Propyl carbinol, n-Butanol, n-Butyl Alcohol

B0944-500ML
TCB0944-500MLEA 44.77 USD
TCB0944-500ML
1-Butanol ≥99.0%
1-Butanol

[for HPLC Solvent] CAS Number: 71-36-3 MDL Number: MFCD00002964 Molecular Formula: C4H10O Molecular Weight: 74.12 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 118 Melting point (°C): -90 Flash Point (°C): 37 Specific Gravity (20/20): 0.81


Product Categories: Analytical Science, Solvents for HPLC Other Product Names / Synonyms: Butyl Alcohol

Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 117.6
Melting Pt: -89
Density: 0.81 (20 °C)
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3 III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfc7eee32d6fd72aa7b407984592c24fada323ec&vid=386ea0a7a055dc8447089989f10d0985d267c805&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR