4-Bromo-1-butene ≥97.0%

Supplier: TCI America

B0920-10G B0920-250G B0920-25G
TCB0920-10GEA 43.73 USD
TCB0920-10G TCB0920-250G TCB0920-25G
4-Bromo-1-butene ≥97.0%
4-Bromo-1-butene

CAS Number: 5162-44-7 MDL Number: MFCD00000258 Molecular Formula: C4H7Br Molecular Weight: 135.00 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 99 Flash Point (°C): 1 Specific Gravity (20/20): 1.34


Other Product Names / Synonyms: 3-Butenyl Bromide

Formula: C₄H₇Br
Boiling Pt: 99 °C
Density: 1.34 (20 °C)
Flash Pt: 1 °C
MDL Number: MFCD00000258
CAS Number: 5162-44-7
UN: 1992
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ae31e7d1df08bb6fb2b1a31b35af04a0112ac51&vid=a21086da25a466913dca81e50703ad7fd2a6b7e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR