N,O-Bis(trimethylsilyl)acetamide kit ≥80% (by GC)

Supplier: TCI America

Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate

B0911-1KIT
TCB0911-KITEA 86.8 USD
TCB0911-KIT
N,O-Bis(trimethylsilyl)acetamide kit ≥80% (by GC)
N,O-Bis(trimethylsilyl)acetamide

Kit BSA 1 mL × 8 / Reaction vial, capacity 2 mL × 8

Formula: C₈H₂₁NOSi₂
MW: 203.43 g/mol
MDL Number: MFCD00008270
CAS Number: 10416-59-8
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=528b38386e346ad13e3d5acd368a10a79554996c&vid=4d42fc44b73663196e202f0a7ade0efd26425507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 75.0% (GC)
Boiling point (°C) 73
Melting point (°C) -24
Flash Point (°C) 42
Specific Gravity (20/20) 0.83
Storage Temperature 0-10°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR