1-Bromo-2-fluorobenzene ≥99.0%

Supplier: TCI America

Synonyms: 2-Bromofluorobenzene

B0883-500G B0883-25G B0883-100G
TCB0883-500GEA 379 USD
TCB0883-500G TCB0883-25G TCB0883-100G
1-Bromo-2-fluorobenzene ≥99.0%
1-Bromo-2-fluorobenzene

CAS Number: 1072-85-1 MDL Number: MFCD00000282 Molecular Formula: C6H4BrF Molecular Weight: 175.00 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 158 Melting point (°C): -8 Flash Point (°C): 48 Specific Gravity (20/20): 1.61


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Benzyne Precursors (Aryne Precursors) Other Product Names / Synonyms: 1-Bromo-2-fluorobenzene

Formula: C₆H₄BrF
MW: 175 g/mol
Boiling Pt: 155
Melting Pt: –8 °C
Density: 1.61 (20 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00000282
CAS Number: 1072-85-1
EINECS: 214-018-3
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8cd08a6c0b62aeb13dd2c9071a815868310cbb97&vid=81f03af473369458c88b09b1e60f10aff9ac1348&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR