9-Bromoanthracene ≥95.0%

Supplier: TCI America

B0872-5G B0872-25G
TCB0872-5GEA 22 USD
TCB0872-5G TCB0872-025G
9-Bromoanthracene ≥95.0%
9-Bromoanthracene

CAS Number: 1564-64-3 MDL Number: MFCD00001243 Molecular Formula: C14H9Br Molecular Weight: 257.13 Purity/Analysis Method: 95.0% (GC) Form: Crystal Melting point (°C): 101

Formula: C₁₄H₉Br
Boiling Pt: 359 °C (1013 hPa)
Melting Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00001243
CAS Number: 1564-64-3
EINECS: 216-359-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a566956ba9da6b8ab581ecaedfcd86e9937c5b6f&vid=864cc8402a971508e9a7a4935630aca7f08be299&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR