1-Butanesulfonyl chloride ≥98.0%

Supplier: TCI America

Synonyms: 1-Butanesulphonylchloride, 1-Butanesulfonylchloride

B0866-500G B0866-25G
TCB0866-500GEA 586.9 USD
TCB0866-500G TCB0866-25G
1-Butanesulfonyl chloride ≥98.0%
1-Butanesulphonyl chloride

CAS Number: 2386-60-9 MDL Number: MFCD00007464 Molecular Formula: C4H9ClO2S Molecular Weight: 156.62 Purity/Analysis Method: 98.0% (T) Form: Clear Liquid Boiling point (°C): 65 Flash Point (°C): 79 Specific Gravity (20/20): 1.22


Other Product Names / Synonyms: Butylsulfonyl Chloride

Formula: C₄H₉ClO₂S
MW: 156.63 g/mol
Boiling Pt: 208
Melting Pt: -25
Density: 1.22 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00007464
CAS Number: 2386-60-9
EINECS: 219-204-8
UN: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e22c4c8f5c27453d18463c6cff08ecbefebed39&vid=5e22c4c8f5c27453d18463c6cff08ecbefebed39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR