2,2'-Butyliminodiethanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

B0725-500ML B0725-25ML
TCB0725-500MLEA 55.94 USD
TCB0725-500ML TCB0725-25ML
2,2'-Butyliminodiethanol ≥98.0% (by GC, titration analysis)
2,2'-Butyliminodiethanol

CAS Number: 102-79-4 MDL Number: MFCD00002856 Molecular Formula: C8H19NO2 Molecular Weight: 161.25 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 275 Melting point (°C): -70 Flash Point (°C): 143 Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: 2,2'-(Butylimino)diethanol, N,N-Bis(2-hydroxyethyl)butylamine

Formula: H₃CCH₂CH₂CH₂N(CH₂CH₂OH)₂
Boiling Pt: 275 °C
Melting Pt: –70 °C
Density: 0.97 (20 °C)
Flash Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00002856
CAS Number: 102-79-4
EINECS: 203-055-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f514b7b787d75cadaf442b4a20c5ab9759b89e2&vid=4f514b7b787d75cadaf442b4a20c5ab9759b89e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR