tert-Butylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

B0709-500ML B0709-25ML B0709-100ML
TCB0709-500MLEA 52.3 USD
TCB0709-500ML TCB0709-25ML TCB0709-100ML
tert-Butylamine ≥98.0% (by GC, titration analysis)
tert-Butylamine

CAS Number: 75-64-9 MDL Number: MFCD00008050 Molecular Formula: C4H11N Molecular Weight: 73.14 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 46 Flash Point (°C): -7 Freezing point (°C): -73 Specific Gravity (20/20): 0.69


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Amines (Multi-component Reaction) Other Product Names / Synonyms: 2-Amino-2-methylpropane

Formula: C₄H₁₁N
Boiling Pt: 46 °C
Melting Pt: –72 °C
Density: 0.69 (20 °C)
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00008050
CAS Number: 75-64-9
EINECS: 200-888-1
UN: 2734
ADR: 8,I
Merck Index: 13,01544
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=558efba197c80a6cf715a9c7543bc31acd41836e&vid=231886ff2ec0f6816d607a672c4fb9c65b4335c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR