tert-Butanol ≥99.0%
Supplier: TCI America
|
Danger
|
CAS Number: 75-65-0
MDL Number: MFCD00004464
Molecular Formula: C4H10O
Molecular Weight: 74.12
Purity/Analysis Method:
Product Categories: Bioscience, Treatment Agents (Staining)
Other Product Names / Synonyms: tert-Butanol, 2-Methyl-2-propanol
Formula:
(CH₃)₃COH Boiling Pt: 82 °C Melting Pt: 25.3 °C Density: 0.7887 g/cm³ (20 °C) Flash Pt: 11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004464 CAS Number: 75-65-0 EINECS: 200-889-7 UN: 1120 ADR: 3,II Merck Index: 12,01577 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...