Brucine sulfate heptahydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
[for Nitrate Analysis]
CAS Number: 60583-39-3
MDL Number: MFCD00013472
Molecular Formula: C23H26N2O4
Molecular Weight: 887.01
Purity/Analysis Method:
Formula:
C₂₃H₂₆N₂O₄ Melting Pt: 180 °C |
MDL Number:
MFCD00013472 CAS Number: 60583-39-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...