Bromothymol blue 0.04% in water

Supplier: TCI America

Synonyms: 3′,3″-Dibromothymolsulphonphthalein, 3′,3″-Dibromothymolsulfonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

B0658-500ML
TCB0658-500MLEA 89.46 USD
TCB0658-500ML
Bromothymol blue 0.04% in water
Bromothymol blue

[for pH Determination]
CAS Number: 76-59-5
MDL Number: MFCD00005872
Molecular Weight: 624.38
Form: Clear Liquid
Color: Brown

Formula: C₂₇H₂₈Br₂O₅S
MW: 624.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005872
CAS Number: 76-59-5
EINECS: 200-971-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=044cede486238cd5b9388b7e9abc3ebcaa13062d&vid=10802eba72013fb7626d7020bfce5422b869e487&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR