2-Bromopyridine ≥98.0%

Supplier: TCI America

B0650-500ML B0650-100ML B0650-25ML
TCB0650-500MLEA 413 USD
TCB0650-500ML TCB0650-100ML TCB0650-25ML
2-Bromopyridine ≥98.0%
2-Bromopyridine

CAS Number: 109-04-6 MDL Number: MFCD00006219 Molecular Formula: C5H4BrN Molecular Weight: 158.00 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 194 Flash Point (°C): 54 Specific Gravity (20/20): 1.63

Formula: C₅H₄BrN
Boiling Pt: 194 °C
Melting Pt: –40.1 °C
Density: 1.63 (20 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00006219
CAS Number: 109-04-6
EINECS: 203-641-6
UN: 2810
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4325252c307a8e2a91e4b2452010866a7fba0556&vid=4325252c307a8e2a91e4b2452010866a7fba0556&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR