2-Bromohexanoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B0601-500G B0601-25G
TCB0601-500GEA 214.61 USD
TCB0601-500G TCB0601-25G
2-Bromohexanoic acid ≥98.0% (by GC, titration analysis)
2-Bromohexanoic acid

CAS Number: 616-05-7 MDL Number: MFCD00004218 Molecular Formula: C6H11BrO2 Molecular Weight: 195.06 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 240 Flash Point (°C): 110 Specific Gravity (20/20): 1.39

Formula: C₆H₁₁BrO₂
Boiling Pt: 240 °C
Density: 1.39 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004218
CAS Number: 616-05-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a153d0e026b978fbbe34d63242d532123c6ae720&vid=a153d0e026b978fbbe34d63242d532123c6ae720&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR