2,5-Dimercapto-1,3,4-thiadiazole ≥95.0%

Supplier: TCI America

Synonyms: 1,3,4-Thiadiazole-2,5-dithiol

B0593-25G B0593-500G
TCB0593-025GEA 30 USD
TCB0593-025G TCB0593-500G
2,5-Dimercapto-1,3,4-thiadiazole ≥95.0%
2,5-Dimercapto-1,3,4-thiadiazole

CAS Number: 1072-71-5 MDL Number: MFCD00003103 Molecular Formula: C2H2N2S3 Molecular Weight: 150.23 Purity/Analysis Method: 95.0% (T) Form: Crystal


Other Product Names / Synonyms: 2,5-Dimercapto-1,3,4-thiadiazole

Formula: C₂H₂N₂S₃
MW: 150.25 g/mol
MDL Number: MFCD00003103
CAS Number: 1072-71-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6959342ba5af67f37f8bbdd94edfc6bcfd25fa0&vid=d6959342ba5af67f37f8bbdd94edfc6bcfd25fa0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR