2-Bromethylamine hydrobromide ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide
B0591-25G
B0591-500G
B0591-100G
TCB0591-025GEA
43
USD
TCB0591-025G
TCB0591-500G
TCB0591-100G
2-Bromethylamine hydrobromide ≥98.0%
2-Bromethylamine hydrobromide
CAS Number: 2576-47-8
MDL Number: MFCD00012886
Molecular Formula: C2H6BrN
Molecular Weight: 204.89
Purity/Analysis Method:
Other Product Names / Synonyms: 2-Aminoethyl Bromide Hydrobromide
Formula:
BrCH₂CH₂NH₃Br MW: 204.89 g/mol Melting Pt: 175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012886 CAS Number: 2576-47-8 EINECS: 219-924-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...