Bromocresol green 0.04% in water

Supplier: TCI America

Synonyms: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

B0579-500ML
TCB0579-500MLEA 66 USD
TCB0579-500ML
Bromocresol green 0.04% in water
Bromocresol green

[for pH Determination]
CAS Number: 76-60-8
MDL Number: MFCD00005874
Molecular Weight: 698.01
Form: Liquid
Color: Green

Formula: C₂₁H₁₄Br₄O₅S
MW: 698.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005874
CAS Number: 76-60-8
EINECS: 200-972-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c3af646275f1ebfc9a2098b52685e1b5751dd21&vid=352c31c2f254c75ad75de0f3781aed4a6d5cece1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR