N,O-Bis(trimethylsilyl)acetamide 25% in acetonitrile

Supplier: TCI America

Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate

B0510-12ML
TCB0510-12MLEA 59 USD
TCB0510-12ML
N,O-Bis(trimethylsilyl)acetamide 25% in acetonitrile
N,O-Bis(trimethylsilyl)acetamide

[Trimethylsilylating Reagent, for NH2 compounds]
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 10
Storage Temperature: 0-10°C


Other Product Names / Synonyms: TMS-BA, BSA

Formula: C₈H₂₁NOSi₂
MW: 203.43 g/mol
Flash Pt: 10 °C
MDL Number: MFCD00008270
CAS Number: 10416-59-8
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1005f7d84336f89772ac9abdb5cf250c3fe65ad&vid=4c01c12c82bc4baef251bed014de3f68f39fbd8d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR