Bis(2-hydroxyethyl)dithiocarbamic acid zinc(II) salt ≥98.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 19163-92-9
MDL Number: MFCD00070508
Molecular Formula: C5H11NO2S2
Molecular Weight: 425.90
Purity/Analysis Method:
Other Product Names / Synonyms: Bis[bis(2-hydroxyethyl)dithiocarbamato]zinc(II)
Formula:
C₅H₁₁NO₂S₂ Melting Pt: 151 °C |
MDL Number:
MFCD00070508 CAS Number: 19163-92-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...