Bicine ≥99.0%
Supplier: TCI America
Synonyms:
2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine
B0484-500G
B0484-25G
B0484-100G
TCB0484-500GEA
264
USD
TCB0484-500G
TCB0484-025G
TCB0484-100G
Bicine ≥99.0%
Bicine
[Good's buffer component for biological research]
CAS Number: 150-25-4
MDL Number: MFCD00004295
Molecular Formula: C6H13NO4
Molecular Weight: 163.17
Purity/Analysis Method:
Product Categories: Bioscience, Good's Buffering Agents
Other Product Names / Synonyms: Bicine, N,N-Bis(2-hydroxyethyl)glycine, Diethanolglycine
Formula:
C₆H₁₃NO₄ Melting Pt: 195 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004295 CAS Number: 150-25-4 EINECS: 205-755-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...