Benzylamine ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
α-Aminotoluene
B0406-500ML
B0406-25ML
TCB0406-500MLEA
80.67
USD
TCB0406-500ML
TCB0406-25ML
Benzylamine ≥99.0%
Benzylamine
CAS Number: 100-46-9
MDL Number: MFCD00008106
Molecular Formula: C7H9N
Molecular Weight: 107.16
Purity/Analysis Method:
Formula:
(C₆H₅)CH₂NH₂ Boiling Pt: 184 °C Melting Pt: 10 °C Density: 0.98 (20 °C) Flash Pt: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008106 CAS Number: 100-46-9 EINECS: 202-854-1 UN: 2735 ADR: 8,II Merck Index: 13,01127 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...