N-Benzylacetamide ≥98.0%
Supplier: TCI America
CAS Number: 588-46-5
MDL Number: MFCD00059204
Molecular Formula: C9H11NO
Molecular Weight: 149.19
Purity/Analysis Method:
Other Product Names / Synonyms: N-Acetylbenzylamine
Formula:
C₉H₁₁NO Boiling Pt: 157 °C Melting Pt: 61 °C |
MDL Number:
MFCD00059204 CAS Number: 588-46-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...