4,4'-Bis(diethylamino)benzophenone ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B0139-25G B0139-500G B0139-100G
TCB0139-025GEA 48 USD
TCB0139-025G TCB0139-500G TCB0139-100G
4,4'-Bis(diethylamino)benzophenone ≥98.0% (by titrimetric analysis)
4,4'-Bis(diethylamino)benzophenone

CAS Number: 90-93-7 MDL Number: MFCD00009044 Molecular Formula: C21H28N2O Molecular Weight: 324.47 Purity/Analysis Method: 98.0% (T) Form: Crystal Melting point (°C): 94 Flash Point (°C): 151 Lambda max.: 352 nm (THF)


Product Categories: Materials Chemistry, Polymerization Initiators Other Product Names / Synonyms: N,N,N',N'-Tetraethyl-4,4'-diaminobenzophenone

Formula: C₂₁H₂₈N₂O
Boiling Pt: 420 °C (1013 hPa)
Melting Pt: 94 °C
Flash Pt: 151 °C
Storage Temperature: Ambient
MDL Number: MFCD00009044
CAS Number: 90-93-7
EINECS: 202-025-4
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c55c245653fd9bc16a4c7a38e7f5dd0ce0d91ae4&vid=c55c245653fd9bc16a4c7a38e7f5dd0ce0d91ae4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR