Saccharin sodium dihydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 6155-57-3
MDL Number: MFCD00151213
Molecular Formula: C7H5NO3S
Molecular Weight: 205.16
Purity/Analysis Method:
Product Categories: Bioscience, Obesity/Diabete Research Reagents
Other Product Names / Synonyms: o-Benzoic Sulfimide Sodium Salt, Saccharin Sodium Salt
Formula:
C₇H₅NNaO₃S·2H₂O Melting Pt: 228 °C |
MDL Number:
MFCD00151213 CAS Number: 6155-57-3 EINECS: 204-886-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...