Bromophenol blue 0.1% in ethanol 50%

Supplier: TCI America
Danger

Synonyms: Bromphenol Blue Sultone Form, 3,3',5,5'-Tetrabromophenolsulfonphthalein, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein, Tetrabromophenol-sulfonephthalein

B0118-500ML
TCB0118-500MLEA 78.72 USD
TCB0118-500ML
Bromophenol blue 0.1% in ethanol 50%
Bromophenol blue

[for Titration]
CAS Number: 115-39-9
MDL Number: MFCD00005875
Molecular Weight: 669.96
Form: Clear Liquid
Color: Yellow Red
Flash Point (°C): 24

Formula: C₁₉H₁₀Br₄O₅S
MW: 669.97 g/mol
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aea6556809d6619210e8e3a9e5846b273b75ee22&vid=827c3831265032450566f693509b28e73c03e80c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR